Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO24
Entrez Gene ID
26261
Gene Name
F-box protein 24
Gene Aliases
FBX24
Chromosome Location
Chr.7: 100582490 - 100601117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163499.1 NP_001156971.1 7 - 8 1103 63
NM_012172.4 NP_036304.2 7 - 8 1253 63
NM_033506.2 NP_277041.1 7 - 8 1364 63
XM_005250259.3 XP_005250316.1 7 - 8 1538 63
XM_011516022.1 XP_011514324.1 7 - 8 1096 63
XM_011516023.2 XP_011514325.1 6 - 7 1355 63
XM_017011961.1 XP_016867450.1 8 - 9 2839 63
GenBank mRNA AK292240.1 - 7 - 8 1371 63
AK301864.1 - 7 - 8 1197 63
AK302252.1 - 7 - 8 1103 63
AL136811.1 - 7 - 8 1309 63
BC067293.1 - 7 - 8 1304 63

View Full Product Details