Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID4A
Entrez Gene ID
5926
Gene Name
AT-rich interaction domain 4A
Gene Aliases
RBBP-1, RBBP1, RBP-1, RBP1
Chromosome Location
Chr.14: 58298385 - 58373733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002892.3 NP_002883.3 23 - 24 3926 105
NM_023000.2 NP_075376.2 23 - 24 3764 105
NM_023001.2 NP_075377.2 22 - 23 3719 105
XM_005267964.2 XP_005268021.1 23 - 24 4105 105
XM_005267965.2 XP_005268022.1 22 - 23 3898 105
XM_017021560.1 XP_016877049.1 23 - 24 4126 105
XM_017021561.1 XP_016877050.1 23 - 24 3903 105
XM_017021562.1 XP_016877051.1 23 - 24 4066 105
XM_017021563.1 XP_016877052.1 22 - 23 3919 105
XM_017021564.1 XP_016877053.1 21 - 22 3734 105
XM_017021565.1 XP_016877054.1 13 - 14 4088 105
GenBank mRNA S57153.1 - 11 - 12 2661 105
S57160.1 - 11 - 12 2499 105
S57162.1 - 10 - 11 2454 105
S66427.1 - 23 - 24 3786 105

View Full Product Details