The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153646.3 | NP_705932.2 | 3 - 4 | 339 | 111 |
NM_153647.3 | NP_705933.2 | 4 - 5 | 412 | 111 | |
NM_153648.3 | NP_705934.1 | 3 - 4 | 241 | 111 | |
XM_005267342.2 | XP_005267399.1 | 2 - 3 | 125 | 111 | |
XM_011536436.2 | XP_011534738.1 | 4 - 5 | 562 | 111 | |
XM_011536437.2 | XP_011534739.1 | 4 - 5 | 562 | 111 | |
XM_011536438.2 | XP_011534740.1 | 4 - 5 | 562 | 111 | |
XM_011536439.2 | XP_011534741.1 | 4 - 5 | 562 | 111 | |
GenBank mRNA | AF520704.1 | - | 3 - 4 | 289 | 111 |
AF520705.1 | - | 3 - 4 | 289 | 111 | |
AF520706.1 | - | 3 - 4 | 241 | 111 | |
AK096171.1 | - | 2 - 3 | 131 | 111 | |
AK295059.1 | - | 4 - 5 | 412 | 111 | |
BC069653.1 | - | 3 - 4 | 289 | 111 |