Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECHDC1
Entrez Gene ID
55862
Gene Name
ethylmalonyl-CoA decarboxylase 1
Gene Aliases
HEL-S-76, MMCD, dJ351K20.2
Chromosome Location
Chr.6: 127288712 - 127343609 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002030.1 NP_001002030.1 4 - 5 686 81
NM_001105544.1 NP_001099014.1 3 - 4 464 81
NM_001139510.1 NP_001132982.1 4 - 5 464 81
XM_005267047.2 XP_005267104.1 5 - 6 611 81
XM_005267048.1 XP_005267105.1 4 - 5 785 81
XM_017011056.1 XP_016866545.1 5 - 6 602 81
GenBank mRNA AK025760.1 - 4 - 5 436 81
AK025796.1 - 4 - 5 464 81
AK303812.1 - 3 - 4 232 81
AL834469.1 - 4 - 5 625 81
BC003549.1 - 4 - 5 648 81
BE896631.1 - 3 - 4 316 81

View Full Product Details