Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
Chromosome Location
Chr.14: 74285277 - 74303064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005050.3 NP_005041.1 13 - 14 1573 94
NR_003256.2 12 - 13 1445 94
XM_005267939.3 XP_005267996.1 13 - 14 1501 94
XM_005267940.3 XP_005267997.1 12 - 13 1380 94
XM_005267941.4 XP_005267998.1 12 - 13 1335 94
XM_005267942.4 XP_005267999.1 11 - 12 1222 94
XM_005267946.3 XP_005268003.1 11 - 12 1217 94
XM_005267949.3 XP_005268006.1 12 - 13 1431 94
XM_006720223.2 XP_006720286.1 12 - 13 1385 94
XM_011537041.2 XP_011535343.1 11 - 12 1243 94
XM_017021531.1 XP_016877020.1 13 - 14 1506 94
XM_017021532.1 XP_016877021.1 12 - 13 1380 94
XM_017021533.1 XP_016877022.1 12 - 13 1380 94
XM_017021534.1 XP_016877023.1 11 - 12 1187 94
XM_017021535.1 XP_016877024.1 12 - 13 1411 94
XM_017021536.1 XP_016877025.1 11 - 12 1231 94
XM_017021537.1 XP_016877026.1 10 - 11 1114 94
XM_017021538.1 XP_016877027.1 11 - 12 1232 94
XM_017021539.1 XP_016877028.1 11 - 12 1215 94
XM_017021540.1 XP_016877029.1 10 - 11 1084 94
XM_017021541.1 XP_016877030.1 11 - 12 1222 94
XM_017021542.1 XP_016877031.1 9 - 10 988 94
XM_017021543.1 XP_016877032.1 10 - 11 1092 94
XM_017021544.1 XP_016877033.1 12 - 13 1411 94
GenBank mRNA AF009746.1 - 13 - 14 1472 94
AK291332.1 - 13 - 14 1443 94
AK297934.1 - 11 - 12 1308 94
AK308382.1 - 12 - 13 1326 94
BC012815.2 - 13 - 14 1427 94
BT007412.1 - 13 - 14 1421 94
CR457104.1 - 13 - 14 1421 94

View Full Product Details