The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 15 - 16 | 2017 | 64 |
XM_005268209.3 | XP_005268266.1 | 15 - 16 | 1821 | 64 | |
XM_017021765.1 | XP_016877254.1 | 15 - 16 | 1784 | 64 | |
GenBank mRNA | AB082526.1 | - | 15 - 16 | 1953 | 64 |
AK128693.1 | - | 15 - 16 | 2421 | 64 | |
AL832633.2 | - | 9 - 10 | 1642 | 64 | |
AY048580.1 | - | 15 - 16 | 1847 | 64 | |
AY080896.1 | - | 15 - 16 | 1859 | 64 | |
BC093881.1 | - | 15 - 16 | 1928 | 64 | |
BC112101.1 | - | 15 - 16 | 1928 | 64 |