Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 5 - 6 765 126
NM_020784.2 NP_065835.2 5 - 6 765 126
XM_017021505.1 XP_016876994.1 5 - 6 745 126
GenBank mRNA AB037765.2 - 5 - 6 677 126
AK097748.1 - 5 - 6 552 126
BC137081.1 - 5 - 6 645 126
BC137082.1 - 5 - 6 645 126
BC142650.1 - 5 - 6 688 126
BC144469.1 - 5 - 6 645 126
BC150205.1 - 5 - 6 686 126
BC152428.1 - 5 - 6 677 126

View Full Product Details