Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
Chromosome Location
Chr.11: 65961689 - 65980136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005146.4 NP_005137.1 16 - 17 2130 56
XM_011545344.1 XP_011543646.1 16 - 17 2159 56
XM_011545345.2 XP_011543647.1 16 - 17 1922 56
GenBank mRNA AB006198.1 - 16 - 17 2076 56
AF353625.1 - 16 - 17 2071 56
AK223016.1 - 16 - 17 2091 56
AK293200.1 - 17 - 18 1997 56
AK298184.1 - 9 - 10 1271 56
BC001058.2 - 16 - 17 2079 56
BC018607.1 - 8 - 9 962 56
BT006637.1 - 16 - 17 2038 56
BX648500.1 - 16 - 17 1880 56
Y14314.1 - 16 - 17 1909 56

View Full Product Details