Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
Chromosome Location
Chr.11: 65961689 - 65980136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005146.4 NP_005137.1 8 - 9 1070 90
XM_011545344.1 XP_011543646.1 8 - 9 1099 90
XM_011545345.2 XP_011543647.1 8 - 9 862 90
GenBank mRNA AB006198.1 - 8 - 9 1016 90
AF353625.1 - 8 - 9 1011 90
AK223016.1 - 8 - 9 1031 90
AK293200.1 - 9 - 10 937 90
BC001058.2 - 8 - 9 1019 90
BT006637.1 - 8 - 9 978 90
BX648500.1 - 8 - 9 820 90
Y14314.1 - 8 - 9 852 90

View Full Product Details