The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182493.2 | NP_872299.2 | 2 - 3 | 687 | 135 |
XM_006721334.3 | XP_006721397.1 | 2 - 3 | 155 | 135 | |
GenBank mRNA | AB451455.1 | - | 2 - 3 | 571 | 135 |
AJ247087.1 | - | 2 - 3 | 687 | 135 | |
BC109097.1 | - | 2 - 3 | 584 | 135 | |
DA561361.1 | - | 2 - 3 | 155 | 135 |