The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307990.1 | NP_001294919.1 | 17 - 18 | 1277 | 89 |
NM_004579.4 | NP_004570.2 | 17 - 18 | 1277 | 89 | |
XM_011545203.2 | XP_011543505.1 | 17 - 18 | 1271 | 89 | |
XM_011545204.2 | XP_011543506.1 | 16 - 17 | 1139 | 89 | |
XM_017018093.1 | XP_016873582.1 | 16 - 17 | 1139 | 89 | |
XM_017018094.1 | XP_016873583.1 | 16 - 17 | 1171 | 89 | |
XM_017018095.1 | XP_016873584.1 | 16 - 17 | 1171 | 89 | |
XM_017018096.1 | XP_016873585.1 | 17 - 18 | 1271 | 89 | |
GenBank mRNA | AK298383.1 | - | 17 - 18 | 1214 | 89 |
BC039839.1 | - | 17 - 18 | 1184 | 89 | |
BC047865.1 | - | 17 - 18 | 1264 | 89 | |
U07349.1 | - | 17 - 18 | 1208 | 89 |