The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307990.1 | NP_001294919.1 | 21 - 22 | 1577 | 57 |
NM_004579.4 | NP_004570.2 | 21 - 22 | 1601 | 57 | |
XM_011545203.2 | XP_011543505.1 | 21 - 22 | 1595 | 57 | |
XM_011545204.2 | XP_011543506.1 | 20 - 21 | 1463 | 57 | |
XM_017018093.1 | XP_016873582.1 | 20 - 21 | 1439 | 57 | |
XM_017018094.1 | XP_016873583.1 | 20 - 21 | 1495 | 57 | |
XM_017018095.1 | XP_016873584.1 | 20 - 21 | 1495 | 57 | |
XM_017018096.1 | XP_016873585.1 | 21 - 22 | 1595 | 57 | |
GenBank mRNA | AK298383.1 | - | 21 - 22 | 1538 | 57 |
BC039839.1 | - | 21 - 22 | 1484 | 57 | |
BC047865.1 | - | 21 - 22 | 1564 | 57 | |
U07349.1 | - | 21 - 22 | 1532 | 57 |