The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282538.1 | NP_001269467.1 | 2 - 3 | 344 | 73 |
NM_002074.4 | NP_002065.1 | 2 - 3 | 344 | 73 | |
XM_017001060.1 | XP_016856549.1 | 2 - 3 | 118 | 73 | |
GenBank mRNA | AK122949.1 | - | 2 - 3 | 128 | 73 |
AK312614.1 | - | 3 - 4 | 105 | 73 | |
BC004186.2 | - | 2 - 3 | 227 | 73 | |
BC008991.1 | - | 2 - 3 | 283 | 73 | |
BC114618.1 | - | 1 - 2 | 66 | 73 | |
BQ881467.1 | - | 2 - 3 | 344 | 73 | |
X04526.1 | - | 2 - 3 | 231 | 73 |