The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017426.3 | NP_059122.2 | 2 - 3 | 286 | 65 |
NR_103780.1 | 2 - 3 | 286 | 65 | ||
NR_103781.1 | 2 - 3 | 286 | 65 | ||
NR_103782.1 | 4 - 5 | 471 | 65 | ||
XM_006714237.1 | XP_006714300.1 | 3 - 4 | 263 | 65 | |
XM_011532033.2 | XP_011530335.1 | 2 - 3 | 229 | 65 | |
XM_017008294.1 | XP_016863783.1 | 2 - 3 | 673 | 65 | |
XM_017008295.1 | XP_016863784.1 | 2 - 3 | 673 | 65 | |
GenBank mRNA | AF157322.1 | - | 2 - 3 | 167 | 65 |
AK001517.1 | - | 2 - 3 | 168 | 65 | |
AK222752.1 | - | 2 - 3 | 170 | 65 | |
AK223352.1 | - | 2 - 3 | 169 | 65 | |
AK299442.1 | - | 2 - 3 | 168 | 65 | |
AK307571.1 | - | 4 - 5 | 361 | 65 | |
AK315160.1 | - | 2 - 3 | 166 | 65 | |
BC012559.1 | - | 2 - 3 | 183 | 65 | |
BX538002.1 | - | 2 - 3 | 186 | 65 | |
DA577078.1 | - | 2 - 3 | 286 | 65 |