The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024430.3 | NP_077748.3 | 10 - 11 | 829 | 110 |
XM_011526253.2 | XP_011524555.1 | 9 - 10 | 794 | 110 | |
XM_011526256.1 | XP_011524558.1 | 9 - 10 | 705 | 110 | |
GenBank mRNA | AK023100.1 | - | 6 - 7 | 397 | 110 |
DA943980.1 | - | 1 - 2 | 87 | 110 | |
KU178456.1 | - | 10 - 11 | 733 | 110 | |
KU178457.1 | - | 9 - 10 | 655 | 110 |