The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000758.3 | NP_000749.2 | 2 - 3 | 226 | 85 |
GenBank mRNA | AF510855.1 | - | 2 - 3 | 143 | 85 |
AY720441.1 | - | 2 - 3 | 203 | 85 | |
BC108724.1 | - | 2 - 3 | 213 | 85 | |
BC113924.1 | - | 2 - 3 | 217 | 85 | |
BC113999.1 | - | 2 - 3 | 217 | 85 | |
KP271009.1 | - | 2 - 3 | 143 | 85 | |
M10663.1 | - | 2 - 3 | 202 | 85 | |
M11220.1 | - | 2 - 3 | 226 | 85 | |
M11734.1 | - | 2 - 3 | 209 | 85 |