The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114094.1 | NP_001107566.1 | 12 - 13 | 1041 | 126 |
NM_001258440.1 | NP_001245369.1 | 13 - 14 | 1050 | 126 | |
NM_001258441.1 | NP_001245370.1 | 12 - 13 | 981 | 126 | |
NM_001258442.1 | NP_001245371.1 | 10 - 11 | 735 | 126 | |
NM_013314.3 | NP_037446.1 | 13 - 14 | 1110 | 126 | |
NR_047680.1 | 12 - 13 | 874 | 126 | ||
NR_047681.1 | 11 - 12 | 822 | 126 | ||
NR_047682.1 | 11 - 12 | 817 | 126 | ||
NR_047683.1 | 11 - 12 | 911 | 126 | ||
XM_011539728.2 | XP_011538030.1 | 13 - 14 | 987 | 126 | |
XM_017016159.1 | XP_016871648.1 | 12 - 13 | 919 | 126 | |
GenBank mRNA | AF068180.1 | - | 13 - 14 | 1085 | 126 |
AF068181.1 | - | 12 - 13 | 1016 | 126 | |
AK027779.1 | - | 13 - 14 | 1045 | 126 | |
AK225546.1 | - | 12 - 13 | 1024 | 126 | |
AM180327.1 | - | 10 - 11 | 617 | 126 | |
AM180332.1 | - | 12 - 13 | 863 | 126 | |
AM180333.1 | - | 12 - 13 | 863 | 126 | |
AM180334.1 | - | 12 - 13 | 863 | 126 | |
AM180335.1 | - | 9 - 10 | 501 | 126 | |
AM180336.1 | - | 13 - 14 | 932 | 126 | |
AM180337.1 | - | 13 - 14 | 932 | 126 | |
AM180338.1 | - | 11 - 12 | 704 | 126 | |
AM180339.1 | - | 11 - 12 | 699 | 126 | |
AM180340.1 | - | 11 - 12 | 793 | 126 | |
AM180342.1 | - | 12 - 13 | 882 | 126 | |
AM180343.1 | - | 12 - 13 | 882 | 126 | |
AM180344.1 | - | 10 - 11 | 772 | 126 | |
AM180345.1 | - | 10 - 11 | 743 | 126 | |
AM180346.1 | - | 10 - 11 | 799 | 126 | |
BC018906.2 | - | 13 - 14 | 1024 | 126 | |
BF975580.1 | - | 1 - 2 | 41 | 126 |