The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000014.4 | NP_000005.2 | 31 - 32 | 4217 | 93 |
XM_006719056.2 | XP_006719119.1 | 31 - 32 | 4195 | 93 | |
GenBank mRNA | AB209614.2 | - | 31 - 32 | 4128 | 93 |
AK307832.1 | - | 30 - 31 | 3954 | 93 | |
AK312482.1 | - | 31 - 32 | 4117 | 93 | |
AY591530.1 | - | 31 - 32 | 4182 | 93 | |
BC026246.1 | - | 13 - 14 | 1845 | 93 | |
BC040071.1 | - | 31 - 32 | 4156 | 93 | |
BX647329.1 | - | 14 - 15 | 1894 | 93 | |
BX648622.1 | - | - | 3601 | 93 | |
CR749334.1 | - | 31 - 32 | 4175 | 93 | |
M11313.1 | - | 31 - 32 | 4147 | 93 | |
M36501.1 | - | 12 - 13 | 1611 | 93 |