Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK4
Entrez Gene ID
9622
Gene Name
kallikrein related peptidase 4
Gene Aliases
AI2A1, ARM1, EMSP, EMSP1, KLK-L1, PRSS17, PSTS, kallikrein
Chromosome Location
Chr.19: 50906352 - 50910745 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004917.4 NP_004908.4 2 - 3 221 59
XM_011527545.2 XP_011525847.1 2 - 3 228 59
XM_011527546.2 XP_011525848.1 2 - 3 228 59
GenBank mRNA AF113140.1 - 2 - 3 221 59
AF126401.1 - 1 - 2 158 59
AF259965.1 - 1 - 2 114 59
AF259966.1 - 1 - 2 74 59
AF259968.1 - 1 - 2 100 59
AF259969.1 - 2 - 3 221 59
AF259970.1 - 1 - 2 74 59
AF259971.1 - 1 - 2 74 59
BC069325.1 - 2 - 3 221 59
BC069403.1 - 2 - 3 221 59
BC069429.1 - 2 - 3 221 59
BC069489.1 - 2 - 3 221 59
BC096175.1 - 2 - 3 221 59
BC096177.3 - 2 - 3 221 59
BC096178.3 - 2 - 3 221 59

View Full Product Details