The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002923.2 | 3 - 4 | 505 | 85 | |
NR_033732.1 | 3 - 4 | 505 | 85 | ||
GenBank mRNA | AK097140.1 | - | 3 - 4 | 505 | 85 |
BC063452.1 | - | 3 - 4 | 488 | 85 | |
BC110520.1 | - | 2 - 3 | 380 | 85 | |
BC110521.1 | - | 2 - 3 | 380 | 85 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | testis-specific transcript, Y-linked 14 (non-protein coding) | TTTY14 | CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14 | -396.4kb | Sense | Chr. - 18872501 - 19077547 on Build GRCh38 |
Downstream | taxilin gamma pseudogene, Y-linked | TXLNGY | CYorf15A, CYorf15B, TXLNG2P | 92.5kb | Anti-sense | Chr. - 19567358 - 19607170 on Build GRCh38 |