The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052957.4 | NP_443189.1 | 9 - 10 | 1963 | 65 |
XM_006724716.3 | XP_006724779.1 | 9 - 10 | 1516 | 65 | |
XM_011531063.2 | XP_011529365.1 | 8 - 9 | 1783 | 65 | |
XM_011531064.1 | XP_011529366.1 | 9 - 10 | 1566 | 65 | |
XM_011531066.2 | XP_011529368.1 | 6 - 7 | 1702 | 65 | |
XM_011531067.2 | XP_011529369.1 | 6 - 7 | 1702 | 65 | |
XM_017029965.1 | XP_016885454.1 | 10 - 11 | 1617 | 65 | |
GenBank mRNA | AJ311392.1 | - | 8 - 9 | 1464 | 65 |
AK097433.1 | - | 9 - 10 | 1577 | 65 | |
BC136259.1 | - | 9 - 10 | 1531 | 65 |