The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052957.4 | NP_443189.1 | 12 - 13 | 2453 | 127 |
XM_006724716.3 | XP_006724779.1 | 12 - 13 | 2006 | 127 | |
XM_011531063.2 | XP_011529365.1 | 11 - 12 | 2273 | 127 | |
XM_011531064.1 | XP_011529366.1 | 12 - 13 | 2056 | 127 | |
XM_011531066.2 | XP_011529368.1 | 9 - 10 | 2192 | 127 | |
XM_011531067.2 | XP_011529369.1 | 9 - 10 | 2192 | 127 | |
XM_017029965.1 | XP_016885454.1 | 13 - 14 | 2107 | 127 | |
GenBank mRNA | AJ311392.1 | - | 11 - 12 | 1954 | 127 |
AK097433.1 | - | 12 - 13 | 2067 | 127 | |
AK127607.1 | - | 11 - 12 | 2489 | 127 | |
BC136259.1 | - | 12 - 13 | 2021 | 127 |