Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location
Chr.12: 1969677 - 2697949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000719.6 NP_000710.5 40 - 41 5269 86
NM_001129827.1 NP_001123299.1 42 - 43 5413 86
NM_001129829.1 NP_001123301.1 40 - 41 5392 86
NM_001129830.2 NP_001123302.2 40 - 41 5234 86
NM_001129831.1 NP_001123303.1 41 - 42 5353 86
NM_001129832.1 NP_001123304.1 41 - 42 5329 86
NM_001129833.1 NP_001123305.1 40 - 41 5326 86
NM_001129834.1 NP_001123306.1 40 - 41 5326 86
NM_001129835.1 NP_001123307.1 40 - 41 5326 86
NM_001129836.1 NP_001123308.1 40 - 41 5320 86
NM_001129837.1 NP_001123309.1 39 - 40 5293 86
NM_001129838.1 NP_001123310.1 39 - 40 5293 86
NM_001129839.1 NP_001123311.1 39 - 40 5287 86
NM_001129840.1 NP_001123312.1 40 - 41 5269 86
NM_001129841.1 NP_001123313.1 40 - 41 5269 86
NM_001129842.1 NP_001123314.1 40 - 41 5269 86
NM_001129843.1 NP_001123315.1 40 - 41 5269 86
NM_001129844.1 NP_001123316.1 40 - 41 5260 86
NM_001129846.1 NP_001123318.1 39 - 40 5236 86
NM_001167623.1 NP_001161095.1 40 - 41 5269 86
NM_001167624.2 NP_001161096.2 40 - 41 5234 86
NM_001167625.1 NP_001161097.1 39 - 40 5236 86
NM_199460.3 NP_955630.3 42 - 43 5378 86
XM_006719017.2 XP_006719080.1 40 - 41 5323 86
XM_011521020.2 XP_011519322.1 41 - 42 5398 86
XM_011521023.2 XP_011519325.1 41 - 42 5583 86
XM_017019926.1 XP_016875415.1 44 - 45 6846 86
XM_017019927.1 XP_016875416.1 44 - 45 6846 86
XM_017019928.1 XP_016875417.1 44 - 45 6846 86
XM_017019929.1 XP_016875418.1 44 - 45 6789 86
XM_017019930.1 XP_016875419.1 44 - 45 6789 86
XM_017019931.1 XP_016875420.1 43 - 44 6705 86
XM_017019932.1 XP_016875421.1 40 - 41 6477 86
XM_017019933.1 XP_016875422.1 42 - 43 6594 86
XM_017019934.1 XP_016875423.1 42 - 43 6585 86
XM_017019935.1 XP_016875424.1 42 - 43 6585 86
XM_017019936.1 XP_016875425.1 41 - 42 6567 86
XM_017019937.1 XP_016875426.1 41 - 42 6567 86
XM_017019938.1 XP_016875427.1 41 - 42 6567 86
XM_017019939.1 XP_016875428.1 41 - 42 6561 86
XM_017019940.1 XP_016875429.1 40 - 41 6534 86
XM_017019941.1 XP_016875430.1 40 - 41 6528 86
XM_017019942.1 XP_016875431.1 41 - 42 6510 86
XM_017019943.1 XP_016875432.1 41 - 42 6510 86
XM_017019944.1 XP_016875433.1 41 - 42 6510 86
XM_017019945.1 XP_016875434.1 41 - 42 6510 86
XM_017019946.1 XP_016875435.1 41 - 42 6510 86
XM_017019947.1 XP_016875436.1 41 - 42 6510 86
XM_017019948.1 XP_016875437.1 41 - 42 6510 86
XM_017019949.1 XP_016875438.1 41 - 42 6510 86
XM_017019950.1 XP_016875439.1 41 - 42 6504 86
XM_017019951.1 XP_016875440.1 41 - 42 6501 86
XM_017019952.1 XP_016875441.1 40 - 41 6477 86
XM_017019953.1 XP_016875442.1 40 - 41 5323 86
XM_017019954.1 XP_016875443.1 40 - 41 5323 86
XM_017019955.1 XP_016875444.1 42 - 43 6585 86
GenBank mRNA AF465484.1 - 40 - 41 5221 86
AJ224873.1 - 40 - 41 4956 86
AK302698.1 - 10 - 11 1311 86
AY830711.1 - 39 - 40 4974 86
AY830712.1 - 40 - 41 5013 86
AY830713.1 - 40 - 41 5079 86
BC146846.1 - 40 - 41 5075 86
L29529.1 - 38 - 39 4855 86
L29534.1 - 39 - 40 4882 86
L29536.1 - 39 - 40 4882 86
Z26294.1 - 9 - 10 1011 86
Z26295.1 - 15 - 16 1740 86
Z34809.1 - 40 - 41 4956 86
Z34810.1 - 40 - 41 4956 86
Z34811.1 - 40 - 41 5013 86
Z34812.1 - 39 - 40 4980 86
Z34813.1 - 39 - 40 4980 86
Z34814.1 - 39 - 40 4923 86
Z34815.1 - 40 - 41 4956 86
Z34816.1 - 40 - 41 4956 86
Z34818.1 - 41 - 42 5016 86
Z34819.1 - 41 - 42 5040 86
Z34820.1 - 40 - 41 5007 86
Z34821.1 - 42 - 43 5100 86
Z34822.1 - 40 - 41 4956 86
Z74996.1 - 40 - 41 5013 86

View Full Product Details