Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location
Chr.12: 1969677 - 2697949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000719.6 NP_000710.5 9 - 10 1701 65
NM_001129827.1 NP_001123299.1 9 - 10 1701 65
NM_001129829.1 NP_001123301.1 9 - 10 1701 65
NM_001129830.2 NP_001123302.2 9 - 10 1666 65
NM_001129831.1 NP_001123303.1 9 - 10 1701 65
NM_001129832.1 NP_001123304.1 9 - 10 1701 65
NM_001129833.1 NP_001123305.1 9 - 10 1701 65
NM_001129834.1 NP_001123306.1 9 - 10 1701 65
NM_001129835.1 NP_001123307.1 9 - 10 1701 65
NM_001129836.1 NP_001123308.1 9 - 10 1701 65
NM_001129837.1 NP_001123309.1 9 - 10 1701 65
NM_001129838.1 NP_001123310.1 9 - 10 1701 65
NM_001129839.1 NP_001123311.1 9 - 10 1701 65
NM_001129840.1 NP_001123312.1 9 - 10 1701 65
NM_001129841.1 NP_001123313.1 9 - 10 1701 65
NM_001129842.1 NP_001123314.1 9 - 10 1701 65
NM_001129843.1 NP_001123315.1 9 - 10 1701 65
NM_001129844.1 NP_001123316.1 9 - 10 1692 65
NM_001129846.1 NP_001123318.1 9 - 10 1701 65
NM_001167623.1 NP_001161095.1 9 - 10 1701 65
NM_001167624.2 NP_001161096.2 9 - 10 1666 65
NM_001167625.1 NP_001161097.1 9 - 10 1701 65
NM_199460.3 NP_955630.3 9 - 10 1666 65
XM_006719017.2 XP_006719080.1 9 - 10 1755 65
XM_017019932.1 XP_016875421.1 10 - 11 2942 65
XM_017019933.1 XP_016875422.1 10 - 11 2942 65
XM_017019936.1 XP_016875425.1 10 - 11 2942 65
XM_017019937.1 XP_016875426.1 10 - 11 2942 65
XM_017019938.1 XP_016875427.1 10 - 11 2942 65
XM_017019939.1 XP_016875428.1 10 - 11 2942 65
XM_017019940.1 XP_016875429.1 10 - 11 2942 65
XM_017019941.1 XP_016875430.1 10 - 11 2942 65
XM_017019942.1 XP_016875431.1 10 - 11 2942 65
XM_017019943.1 XP_016875432.1 10 - 11 2942 65
XM_017019944.1 XP_016875433.1 10 - 11 2942 65
XM_017019945.1 XP_016875434.1 10 - 11 2942 65
XM_017019946.1 XP_016875435.1 10 - 11 2942 65
XM_017019947.1 XP_016875436.1 10 - 11 2942 65
XM_017019948.1 XP_016875437.1 10 - 11 2942 65
XM_017019949.1 XP_016875438.1 10 - 11 2942 65
XM_017019950.1 XP_016875439.1 10 - 11 2942 65
XM_017019951.1 XP_016875440.1 10 - 11 2933 65
XM_017019952.1 XP_016875441.1 10 - 11 2942 65
XM_017019953.1 XP_016875442.1 9 - 10 1755 65
XM_017019954.1 XP_016875443.1 9 - 10 1755 65
GenBank mRNA AF465484.1 - 9 - 10 1653 65
AJ224873.1 - 9 - 10 1388 65
AJ537510.1 - 1 - 2 59 65
AY562396.1 - 2 - 3 275 65
AY830711.1 - 9 - 10 1388 65
AY830712.1 - 9 - 10 1388 65
AY830713.1 - 9 - 10 1388 65
BC146846.1 - 9 - 10 1507 65
L29529.1 - 8 - 9 1320 65
L29534.1 - 8 - 9 1314 65
L29536.1 - 8 - 9 1314 65
Z34809.1 - 9 - 10 1388 65
Z34810.1 - 9 - 10 1388 65
Z34811.1 - 9 - 10 1388 65
Z34812.1 - 9 - 10 1388 65
Z34813.1 - 9 - 10 1388 65
Z34814.1 - 9 - 10 1388 65
Z34815.1 - 9 - 10 1388 65
Z34816.1 - 9 - 10 1388 65
Z34817.1 - 9 - 10 1388 65
Z34818.1 - 9 - 10 1388 65
Z34819.1 - 9 - 10 1388 65
Z34820.1 - 9 - 10 1388 65
Z34821.1 - 9 - 10 1388 65
Z34822.1 - 9 - 10 1388 65
Z74996.1 - 9 - 10 1388 65

View Full Product Details