The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004041.4 | NP_004032.2 | 4 - 5 | 376 | 80 |
NM_020251.3 | NP_064647.1 | 4 - 5 | 376 | 80 | |
XM_011545034.2 | XP_011543336.1 | 5 - 6 | 260 | 80 | |
XM_011545035.2 | XP_011543337.1 | 5 - 6 | 260 | 80 | |
XM_017017750.1 | XP_016873239.1 | 5 - 6 | 260 | 80 | |
XM_017017751.1 | XP_016873240.1 | 5 - 6 | 260 | 80 | |
XM_017017752.1 | XP_016873241.1 | 4 - 5 | 216 | 80 | |
XM_017017753.1 | XP_016873242.1 | 4 - 5 | 280 | 80 | |
XM_017017754.1 | XP_016873243.1 | 4 - 5 | 216 | 80 | |
GenBank mRNA | AF084040.1 | - | 4 - 5 | 152 | 80 |
AF084940.1 | - | 4 - 5 | 152 | 80 | |
BC003636.2 | - | 4 - 5 | 229 | 80 | |
BQ883950.1 | - | 4 - 5 | 399 | 80 | |
FJ348262.1 | - | 4 - 5 | 152 | 80 | |
L04685.1 | - | 4 - 5 | 152 | 80 |