The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099855.1 | NP_001093325.1 | 24 - 25 | 2784 | 102 |
NM_001171876.1 | NP_001165347.1 | 25 - 26 | 2832 | 102 | |
NM_001171877.1 | NP_001165348.1 | 20 - 21 | 2649 | 102 | |
NM_001171878.1 | NP_001165349.1 | 21 - 22 | 2766 | 102 | |
NM_001171879.1 | NP_001165350.1 | 22 - 23 | 2814 | 102 | |
NM_005369.4 | NP_005360.3 | 21 - 22 | 2766 | 102 | |
XM_005262413.4 | XP_005262470.1 | 26 - 27 | 2980 | 102 | |
XM_011531339.2 | XP_011529641.1 | 26 - 27 | 3039 | 102 | |
XM_017029529.1 | XP_016885018.1 | 25 - 26 | 2801 | 102 | |
XM_017029530.1 | XP_016885019.1 | 25 - 26 | 2801 | 102 | |
XM_017029531.1 | XP_016885020.1 | 25 - 26 | 2734 | 102 | |
XM_017029532.1 | XP_016885021.1 | 22 - 23 | 2891 | 102 | |
GenBank mRNA | AB085901.1 | - | 21 - 22 | 2443 | 102 |
AB085902.1 | - | 22 - 23 | 2491 | 102 | |
AK296488.1 | - | 20 - 21 | 2649 | 102 | |
AK296529.1 | - | 24 - 25 | 2884 | 102 | |
AK302957.1 | - | 26 - 27 | 2780 | 102 | |
AK313900.1 | - | 21 - 22 | 2626 | 102 | |
AK316416.1 | - | 25 - 26 | 2820 | 102 | |
AL117234.1 | - | 23 - 24 | 2667 | 102 | |
J03639.1 | - | - | 1499 | 102 | |
X12556.1 | - | 21 - 22 | 2617 | 102 | |
X13230.1 | - | - | 1371 | 102 |