Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
Chromosome Location
Chr.X: 118345997 - 118449961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184966.1 NP_001171895.1 15 - 16 2547 112
NM_019045.4 NP_061918.3 17 - 18 2814 112
XM_011531353.2 XP_011529655.1 16 - 17 2815 112
XM_017029600.1 XP_016885089.1 16 - 17 2747 112
GenBank mRNA AK092077.1 - 14 - 15 1818 112
AK127556.1 - 17 - 18 2725 112
AK299711.1 - 15 - 16 2483 112
AK304432.1 - 17 - 18 2341 112
AL834210.1 - 14 - 15 2081 112
BC028697.2 - 17 - 18 2750 112
BX537578.1 - 17 - 18 2788 112
BX538087.1 - 17 - 18 2756 112
BX648150.1 - 17 - 18 2753 112

View Full Product Details