Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX27
Entrez Gene ID
81609
Gene Name
sorting nexin family member 27
Gene Aliases
MRT1, MY014
Chromosome Location
Chr.1: 151612027 - 151699083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030918.5 NP_112180.4 11 - 12 1585 80
XM_005245511.4 XP_005245568.1 11 - 12 1613 80
XM_017002417.1 XP_016857906.1 10 - 11 1286 80
GenBank mRNA AB007957.2 - 11 - 12 1554 80
AF060509.1 - 5 - 6 526 80
BC012184.1 - 11 - 12 1575 80
BC051817.1 - 8 - 9 834 80
BC100998.1 - 11 - 12 1585 80
BC100999.1 - 11 - 12 1583 80
BC101000.1 - 11 - 12 1585 80
BC101822.1 - 11 - 12 1620 80
BC101824.1 - 11 - 12 1620 80

View Full Product Details