Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX27
Entrez Gene ID
81609
Gene Name
sorting nexin family member 27
Gene Aliases
MRT1, MY014
Chromosome Location
Chr.1: 151612027 - 151699083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030918.5 NP_112180.4 3 - 4 755 116
XM_005245509.2 XP_005245566.1 3 - 4 877 116
XM_005245510.3 XP_005245567.1 3 - 4 487 116
XM_005245511.4 XP_005245568.1 3 - 4 783 116
XM_011510024.2 XP_011508326.1 2 - 3 598 116
XM_011510025.2 XP_011508327.1 2 - 3 456 116
XM_011510026.2 XP_011508328.1 3 - 4 851 116
XM_017002417.1 XP_016857906.1 2 - 3 456 116
GenBank mRNA AB007957.2 - 3 - 4 724 116
AY044866.1 - 2 - 3 460 116
BC012184.1 - 3 - 4 745 116
BC071825.1 - 2 - 3 218 116
BC100998.1 - 3 - 4 755 116
BC100999.1 - 3 - 4 753 116
BC101000.1 - 3 - 4 755 116
BC101822.1 - 3 - 4 790 116
BC101824.1 - 3 - 4 790 116
BC107862.1 - 1 - 2 211 116

View Full Product Details