Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX27
Entrez Gene ID
81609
Gene Name
sorting nexin family member 27
Gene Aliases
MRT1, MY014
Chromosome Location
Chr.1: 151612027 - 151699083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030918.5 NP_112180.4 5 - 6 920 86
XM_005245509.2 XP_005245566.1 5 - 6 1042 86
XM_005245510.3 XP_005245567.1 5 - 6 652 86
XM_005245511.4 XP_005245568.1 5 - 6 948 86
XM_011510024.2 XP_011508326.1 4 - 5 763 86
XM_011510025.2 XP_011508327.1 4 - 5 621 86
XM_011510026.2 XP_011508328.1 5 - 6 1016 86
XM_017002417.1 XP_016857906.1 4 - 5 621 86
GenBank mRNA AB007957.2 - 5 - 6 889 86
AY044866.1 - 4 - 5 625 86
BC012184.1 - 5 - 6 910 86
BC051817.1 - 2 - 3 169 86
BC071825.1 - 4 - 5 383 86
BC100998.1 - 5 - 6 920 86
BC100999.1 - 5 - 6 918 86
BC101000.1 - 5 - 6 920 86
BC101822.1 - 5 - 6 955 86
BC101824.1 - 5 - 6 955 86
BC107862.1 - 3 - 4 376 86

View Full Product Details