Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
Chromosome Location
Chr.1: 150996737 - 151008378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040217.2 NP_001035307.1 8 - 9 1473 82
NM_001163258.1 NP_001156730.1 9 - 10 1536 82
NM_001163259.1 NP_001156731.1 8 - 9 1438 82
NM_001163260.1 NP_001156732.1 8 - 9 1349 82
NM_001319998.1 NP_001306927.1 9 - 10 2853 82
NM_018379.4 NP_060849.2 9 - 10 2137 82
XM_005245321.3 XP_005245378.1 8 - 9 2114 82
XM_005245325.3 XP_005245382.1 8 - 9 1469 82
XM_005245327.3 XP_005245384.1 7 - 8 1310 82
XM_005245328.3 XP_005245385.1 7 - 8 1307 82
XM_011509759.2 XP_011508061.1 7 - 8 1690 82
XM_017001770.1 XP_016857259.1 8 - 9 2111 82
XM_017001771.1 XP_016857260.1 8 - 9 1849 82
XM_017001772.1 XP_016857261.1 8 - 9 1849 82
XM_017001775.1 XP_016857264.1 7 - 8 1952 82
XM_017001777.1 XP_016857266.1 8 - 9 1463 82
GenBank mRNA AB037811.1 - 7 - 8 1322 82
AK002142.1 - 8 - 9 1199 82
AK125493.1 - 8 - 9 1173 82
AK125959.1 - 9 - 10 2853 82
AK301946.1 - 9 - 10 1536 82
AK303962.1 - 8 - 9 1126 82
BC032321.1 - 8 - 9 1613 82
BC075812.1 - 9 - 10 2124 82

View Full Product Details