Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SVIL
Entrez Gene ID
6840
Gene Name
supervillin
Gene Aliases
-
Chromosome Location
Chr.10: 29457338 - 29737001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323599.1 NP_001310528.1 35 - 36 5918 83
NM_001323600.1 NP_001310529.1 33 - 34 5519 83
NM_003174.3 NP_003165.2 32 - 33 5304 83
NM_021738.2 NP_068506.2 34 - 35 6885 83
XM_005252570.2 XP_005252627.1 38 - 39 6908 83
XM_005252571.3 XP_005252628.1 38 - 39 6834 83
XM_005252573.2 XP_005252630.1 36 - 37 7040 83
XM_011519630.2 XP_011517932.2 35 - 36 7675 83
XM_011519631.2 XP_011517933.2 35 - 36 7657 83
XM_011519632.2 XP_011517934.2 35 - 36 7582 83
XM_011519633.2 XP_011517935.2 33 - 34 7531 83
XM_011519634.1 XP_011517936.1 38 - 39 6966 83
XM_011519635.1 XP_011517937.1 36 - 37 6933 83
XM_011519636.2 XP_011517938.2 35 - 36 6823 83
XM_011519637.2 XP_011517939.2 33 - 34 6571 83
XM_011519638.2 XP_011517940.2 32 - 33 6475 83
XM_017016575.1 XP_016872064.1 36 - 37 7753 83
XM_017016576.1 XP_016872065.1 35 - 36 7672 83
XM_017016577.1 XP_016872066.1 34 - 35 7627 83
XM_017016578.1 XP_016872067.1 34 - 35 6727 83
XM_017016579.1 XP_016872068.1 33 - 34 6697 83
XM_017016580.1 XP_016872069.1 34 - 35 6652 83
XM_017016581.1 XP_016872070.1 31 - 32 6445 83
XM_017016582.1 XP_016872071.1 30 - 31 6349 83
GenBank mRNA AF051850.1 - 32 - 33 5304 83
AF051851.1 - 28 - 29 4954 83
AF109135.1 - 34 - 35 6885 83
AK304720.1 - 20 - 21 2910 83
AK304799.1 - 18 - 19 2668 83
AL117410.1 - 17 - 18 2690 83
JX467682.1 - 29 - 30 6036 83

View Full Product Details