The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076786.2 | NP_001070254.1 | 4 - 5 | 4128 | 93 |
XM_017018327.1 | XP_016873816.1 | 4 - 5 | 4480 | 93 | |
XM_017018328.1 | XP_016873817.1 | 4 - 5 | 4480 | 93 | |
XM_017018329.1 | XP_016873818.1 | 2 - 3 | 4933 | 93 | |
XM_017018330.1 | XP_016873819.1 | 3 - 4 | 3878 | 93 | |
GenBank mRNA | AK025577.1 | - | 1 - 2 | 855 | 93 |
AK123858.1 | - | 1 - 2 | 2190 | 93 | |
AK125902.1 | - | 1 - 2 | 1676 | 93 | |
AK126023.1 | - | 5 - 6 | 3432 | 93 | |
AL834141.1 | - | 1 - 2 | 2824 | 93 |