Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
Chromosome Location
Chr.11: 43880807 - 43920275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139178.3 NP_631917.1 1 - 2 380 87
GenBank mRNA AB042029.1 - 1 - 2 343 87
AK315266.1 - 1 - 2 376 87
BC067257.1 - 1 - 2 313 87
BC103812.1 - 1 - 2 118 87
BC103813.1 - 1 - 2 118 87
BC103814.2 - 1 - 2 118 87

View Full Product Details