Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
MUC17 | NM_001040105.1 | 3 | 11559 | NP_001035194.1 |
NR_133665.1 | 3 | 11559 | ||
AF016692.1 | 1 | 1116 | AAB71685.1 | |
AF430017.1 | 1 | 1116 | AAL89737.1 | |
AJ606307.1 | 3 | 11559 | CAE54435.1 | |
AJ606308.1 | 3 | 11559 | CAE54436.1 | |
AK026404.1 | 1 | 230 | ||
BC126315.1 | 1 | 1701 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5873 | Chr7:101040374 - 101074468 on Build GRCh38 | Insertion | MUC17 |
nsv428181 | Chr7:100962999 - 101105965 on Build GRCh38 | Gain+Loss | MUC12 MUC17 TRIM56 LOC102724094 MUC3A |
esv2759547 | Chr7:100757867 - 101105965 on Build GRCh38 | Gain+Loss | ZAN ACHE MUC12 SLC12A9 MUC3A MUC17 TRIM56 UFSP1 SRRT LOC102724094 MIR6875 EPHB4 TRIP6 |
esv2734888 | Chr7:99975623 - 101290285 on Build GRCh38 | Deletion | MIR93 ZASP AZGP1 MOGAT3 PLOD3 STAG3L5P C7orf43 MUC17 MBLAC1 MCM7 PILRA PCOLCE-AS1 EPHB4 LOC105375429 ZCWPW1 PMS2P1 PVRIG STAG3L5P-PVRIG2P-PILRB PVRIG2P MUC3A GNB2 LRCH4 GPC2 CLDN15 COPS6 LOC102724094 FIS1 ZNHIT1 GIGYF1 MIR25 AP4M1 MIR106B TRIP6 GATS ACHE TFR2 SLC12A9 EPO NAT16 MIR4658 ZKSCAN1 ZNF3 SPDYE3 TAF6 ZSCAN21 MIR4653 FBXO24 STAG3 AP1S1 MIR6840 MEPCE LAMTOR4 ZAN MUC12 CNPY4 MOSPD3 PILRB SERPINE1 AGFG2 SAP25 GAL3ST4 ACTL6B TSC22D4 TRIM56 UFSP1 SRRT PCOLCE POP7 AZGP1P1 MIR6875 VGF C7orf61 PPP1R35 NYAP1 |
nsv820525 | Chr7:101030087 - 101043704 on Build GRCh38 | Duplication | MUC17 |
nsv1033081 | Chr7:101026348 - 101128543 on Build GRCh38 | Gain | MUC17 TRIM56 SERPINE1 |
nsv1128954 | Chr7:101032019 - 101043819 on Build GRCh38 | Duplication | MUC17 |
nsv1078027 | Chr7:101038318 - 101043719 on Build GRCh38 | Duplication | MUC17 |
nsv1147714 | Chr7:101034218 - 101042919 on Build GRCh38 | Duplication | MUC17 |
nsv831074 | Chr7:100963006 - 101102251 on Build GRCh38 | Loss | MUC12 MUC17 TRIM56 LOC102724094 MUC3A |