Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RORA
Entrez Gene ID
6095
Gene Name
RAR related orphan receptor A
Gene Aliases
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA
Chromosome Location
Chr.15: 60488284 - 61229303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002943.3 NP_002934.1 4 - 5 656 81
NM_134260.2 NP_599022.1 5 - 6 680 81
NM_134261.2 NP_599023.1 4 - 5 510 81
NM_134262.2 NP_599024.1 3 - 4 351 81
XM_005254584.4 XP_005254641.1 4 - 5 4971 81
XM_011521874.1 XP_011520176.1 3 - 4 548 81
XM_011521875.1 XP_011520177.1 4 - 5 380 81
XM_011521877.2 XP_011520179.1 3 - 4 583 81
XM_011521878.1 XP_011520180.1 5 - 6 422 81
XM_017022466.1 XP_016877955.1 3 - 4 2372 81
XM_017022467.1 XP_016877956.1 3 - 4 2045 81
GenBank mRNA AB307694.1 - 4 - 5 425 81
BC008831.2 - 4 - 5 433 81
BC100987.2 - 3 - 4 301 81
BC100988.1 - 3 - 4 301 81
BC100989.1 - 3 - 4 301 81
BC100990.2 - 3 - 4 301 81
HQ692818.1 - 3 - 4 260 81
HQ709174.1 - 3 - 4 260 81
L14611.1 - 3 - 4 281 81
U04897.1 - 4 - 5 526 81
U04898.1 - 5 - 6 613 81
U04899.1 - 4 - 5 578 81

View Full Product Details