The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048200.2 | NP_001041665.1 | 4 - 5 | 1907 | 120 |
NM_001278162.1 | NP_001265091.1 | 4 - 5 | 1699 | 120 | |
NM_001278163.1 | NP_001265092.1 | 4 - 5 | 1420 | 120 | |
NM_005734.4 | NP_005725.3 | 4 - 5 | 1907 | 120 | |
XM_005252729.2 | XP_005252786.1 | 4 - 5 | 1694 | 120 | |
XM_017017076.1 | XP_016872565.1 | 3 - 4 | 1341 | 120 | |
XM_017017077.1 | XP_016872566.1 | 4 - 5 | 1364 | 120 | |
GenBank mRNA | AF004849.1 | - | 3 - 4 | 1346 | 120 |
AF305239.1 | - | 4 - 5 | 1370 | 120 | |
AK293009.1 | - | 4 - 5 | 1700 | 120 | |
AK315277.1 | - | 4 - 5 | 1420 | 120 | |
AY425958.1 | - | 3 - 4 | 259 | 120 |