Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLPH3L
Entrez Gene ID
55204
Gene Name
golgi phosphoprotein 3 like
Gene Aliases
GPP34R
Chromosome Location
Chr.1: 150646225 - 150697196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018178.5 NP_060648.2 1 - 2 200 68
GenBank mRNA AK300635.1 - 1 - 2 158 68
AK315594.1 - 1 - 2 167 68
BI550986.1 - - 166 68
DA513375.1 - 1 - 2 200 68

View Full Product Details