The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018178.5 | NP_060648.2 | 2 - 3 | 394 | 140 |
XM_006711428.1 | XP_006711491.1 | 2 - 3 | 321 | 140 | |
GenBank mRNA | AJ296153.1 | - | 2 - 3 | 222 | 140 |
AK315594.1 | - | 2 - 3 | 361 | 140 | |
BC013870.2 | - | 1 - 2 | 193 | 140 | |
DA513375.1 | - | 2 - 3 | 394 | 140 |