The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206833.1 | NP_001193762.1 | 1 - 2 | 442 | 69 |
NM_006388.3 | NP_006379.2 | 2 - 3 | 343 | 69 | |
NM_182709.2 | NP_874368.1 | 2 - 3 | 343 | 69 | |
NM_182710.2 | NP_874369.1 | 1 - 2 | 442 | 69 | |
XM_006718421.2 | XP_006718484.1 | 2 - 3 | 349 | 69 | |
GenBank mRNA | AK225100.1 | - | 2 - 3 | 292 | 69 |
AK226002.1 | - | 2 - 3 | 292 | 69 | |
AK304664.1 | - | 1 - 2 | 192 | 69 | |
AK313297.1 | - | 2 - 3 | 319 | 69 | |
BC000166.3 | - | 1 - 2 | 65 | 69 | |
BC013211.1 | - | 1 - 2 | 160 | 69 | |
BC064912.1 | - | 2 - 3 | 83 | 69 | |
BC093032.1 | - | 2 - 3 | 110 | 69 | |
BC117167.1 | - | 2 - 3 | 130 | 69 | |
BC143296.1 | - | 2 - 3 | 127 | 69 | |
BG720537.1 | - | 2 - 3 | 334 | 69 | |
BI463394.1 | - | 1 - 2 | 200 | 69 | |
DA043042.1 | - | 2 - 3 | 332 | 69 | |
DB003934.1 | - | 2 - 3 | 343 | 69 | |
DB103287.1 | - | 1 - 2 | 409 | 69 | |
U40989.1 | - | 1 - 2 | 48 | 69 | |
U67734.1 | - | 2 - 3 | 94 | 69 | |
U74667.1 | - | 2 - 3 | 318 | 69 |