The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281428.1 | NP_001268357.1 | 11 - 12 | 1286 | 103 |
NM_012144.3 | NP_036276.1 | 11 - 12 | 1274 | 103 | |
XM_006716758.3 | XP_006716821.1 | 10 - 11 | 917 | 103 | |
XM_011517846.1 | XP_011516148.1 | 11 - 12 | 1286 | 103 | |
XM_011517847.2 | XP_011516149.1 | 11 - 12 | 1286 | 103 | |
XM_011517848.1 | XP_011516150.1 | 11 - 12 | 1286 | 103 | |
XM_011517849.1 | XP_011516151.1 | 11 - 12 | 1286 | 103 | |
XM_011517850.2 | XP_011516152.1 | 11 - 12 | 1286 | 103 | |
XM_017014625.1 | XP_016870114.1 | 11 - 12 | 1274 | 103 | |
GenBank mRNA | AF091619.1 | - | 11 - 12 | 1191 | 103 |
AK290696.1 | - | 11 - 12 | 1198 | 103 | |
AK302186.1 | - | 8 - 9 | 817 | 103 | |
AK314639.1 | - | 11 - 12 | 1186 | 103 | |
BC030583.2 | - | 11 - 12 | 1202 | 103 |