The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012144.3 | NP_036276.1 | 5 - 6 | 638 | 84 |
XM_017014625.1 | XP_016870114.1 | 5 - 6 | 638 | 84 | |
GenBank mRNA | AF091619.1 | - | 5 - 6 | 555 | 84 |
AK290696.1 | - | 5 - 6 | 562 | 84 | |
AK314639.1 | - | 5 - 6 | 550 | 84 | |
BC030583.2 | - | 5 - 6 | 566 | 84 | |
BM013886.1 | - | 5 - 6 | 578 | 84 |