The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289119.1 | NP_001276048.1 | 3 - 4 | 831 | 108 |
NM_001289120.1 | NP_001276049.1 | 3 - 4 | 854 | 108 | |
NM_001289121.1 | NP_001276050.1 | 3 - 4 | 972 | 108 | |
NM_032226.2 | NP_115602.2 | 3 - 4 | 827 | 108 | |
XM_005251608.4 | XP_005251665.1 | 3 - 4 | 1736 | 108 | |
XM_005251612.2 | XP_005251669.1 | 3 - 4 | 794 | 108 | |
XM_011518050.1 | XP_011516352.1 | 3 - 4 | 822 | 108 | |
XM_017015197.1 | XP_016870686.1 | 3 - 4 | 949 | 108 | |
XM_017015198.1 | XP_016870687.1 | 3 - 4 | 1435 | 108 | |
GenBank mRNA | AK026264.1 | - | 3 - 4 | 717 | 108 |
AK074608.1 | - | 2 - 3 | 316 | 108 | |
AK315128.1 | - | 3 - 4 | 844 | 108 | |
BC005990.1 | - | 3 - 4 | 760 | 108 | |
BC036940.1 | - | 3 - 4 | 945 | 108 |