The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031924.4 | NP_114130.3 | 5 - 6 | 1312 | 59 |
XM_005267153.4 | XP_005267210.1 | 3 - 4 | 1290 | 59 | |
XM_017011347.1 | XP_016866836.1 | 4 - 5 | 633 | 59 | |
GenBank mRNA | AF353618.1 | - | 5 - 6 | 911 | 59 |
AK057655.1 | - | 3 - 4 | 639 | 59 | |
AK057931.1 | - | 5 - 6 | 1333 | 59 | |
BC050604.1 | - | 5 - 6 | 1312 | 59 |