The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000381.3 | NP_000372.1 | 6 - 7 | 1549 | 103 |
NM_001098624.2 | NP_001092094.1 | 6 - 7 | 1479 | 103 | |
NM_001193277.1 | NP_001180206.1 | 6 - 7 | 1338 | 103 | |
NM_001193278.1 | NP_001180207.1 | 6 - 7 | 1357 | 103 | |
NM_001193279.1 | NP_001180208.1 | 5 - 6 | 1204 | 103 | |
NM_001193280.1 | NP_001180209.1 | 5 - 6 | 1090 | 103 | |
NM_033289.1 | NP_150631.1 | 6 - 7 | 1435 | 103 | |
NM_033290.3 | NP_150632.1 | 6 - 7 | 1341 | 103 | |
XM_005274536.1 | XP_005274593.1 | 7 - 8 | 1514 | 103 | |
XM_005274537.1 | XP_005274594.1 | 7 - 8 | 1491 | 103 | |
XM_006724492.3 | XP_006724555.1 | 7 - 8 | 2113 | 103 | |
XM_011545525.2 | XP_011543827.1 | 9 - 10 | 1815 | 103 | |
XM_011545526.2 | XP_011543828.1 | 9 - 10 | 1850 | 103 | |
XM_011545527.2 | XP_011543829.1 | 7 - 8 | 1760 | 103 | |
XM_017029534.1 | XP_016885023.1 | 7 - 8 | 1680 | 103 | |
XM_017029535.1 | XP_016885024.1 | 9 - 10 | 1701 | 103 | |
XM_017029536.1 | XP_016885025.1 | 8 - 9 | 1777 | 103 | |
XM_017029537.1 | XP_016885026.1 | 8 - 9 | 1548 | 103 | |
XM_017029538.1 | XP_016885027.1 | 6 - 7 | 1247 | 103 | |
XM_017029539.1 | XP_016885028.1 | 6 - 7 | 1409 | 103 | |
XM_017029540.1 | XP_016885029.1 | 8 - 9 | 1662 | 103 | |
XM_017029541.1 | XP_016885030.1 | 9 - 10 | 1815 | 103 | |
XM_017029542.1 | XP_016885031.1 | 8 - 9 | 1662 | 103 | |
GenBank mRNA | AF035360.1 | - | 6 - 7 | 1235 | 103 |
AF041206.1 | - | 6 - 7 | 1429 | 103 | |
AF041208.1 | - | 6 - 7 | 1549 | 103 | |
AF041210.1 | - | 5 - 6 | 1038 | 103 | |
AF230976.1 | - | 6 - 7 | 1334 | 103 | |
AF230977.1 | - | 6 - 7 | 1334 | 103 | |
AF269101.1 | - | 7 - 8 | 1457 | 103 | |
AK025187.1 | - | 5 - 6 | 519 | 103 | |
AK226104.1 | - | 6 - 7 | 1253 | 103 | |
AK291215.1 | - | 6 - 7 | 1431 | 103 | |
AK297205.1 | - | 7 - 8 | 1345 | 103 | |
AK299061.1 | - | 6 - 7 | 1374 | 103 | |
AK315095.1 | - | 6 - 7 | 1380 | 103 | |
AY539962.1 | - | 5 - 6 | 531 | 103 | |
AY539983.1 | - | 5 - 6 | 574 | 103 | |
AY539994.1 | - | 6 - 7 | 1301 | 103 | |
AY539997.1 | - | 4 - 5 | 448 | 103 | |
AY540009.1 | - | 2 - 3 | 218 | 103 | |
AY540010.1 | - | 2 - 3 | 323 | 103 | |
AY540011.1 | - | 1 - 2 | 280 | 103 | |
AY540012.1 | - | 2 - 3 | 218 | 103 | |
AY540013.1 | - | 1 - 2 | 546 | 103 | |
AY540022.1 | - | 5 - 6 | 1148 | 103 | |
AY540023.1 | - | 5 - 6 | 1034 | 103 | |
BC053626.1 | - | 6 - 7 | 1406 | 103 | |
Y13667.1 | - | 6 - 7 | 1334 | 103 |