The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270419.1 | NP_001257348.1 | 7 - 8 | 920 | 161 |
NM_181581.2 | NP_853559.1 | 7 - 8 | 1070 | 161 | |
NR_073002.1 | 7 - 8 | 1170 | 161 | ||
NR_073003.1 | 6 - 7 | 952 | 161 | ||
NR_073004.1 | 6 - 7 | 909 | 161 | ||
NR_073005.1 | 6 - 7 | 802 | 161 | ||
XM_005250125.4 | XP_005250182.1 | 6 - 7 | 932 | 161 | |
XM_006715844.2 | XP_006715907.1 | 5 - 6 | 808 | 161 | |
XM_006715845.3 | XP_006715908.1 | 4 - 5 | 719 | 161 | |
XM_011515766.2 | XP_011514068.1 | 7 - 8 | 1064 | 161 | |
XM_011515767.2 | XP_011514069.1 | 6 - 7 | 814 | 161 | |
XM_017011708.1 | XP_016867197.1 | 7 - 8 | 1085 | 161 | |
XM_017011709.1 | XP_016867198.1 | 7 - 8 | 1065 | 161 | |
XM_017011710.1 | XP_016867199.1 | 7 - 8 | 919 | 161 | |
XM_017011711.1 | XP_016867200.1 | 6 - 7 | 847 | 161 | |
XM_017011712.1 | XP_016867201.1 | 6 - 7 | 831 | 161 | |
XM_017011713.1 | XP_016867202.1 | 6 - 7 | 901 | 161 | |
GenBank mRNA | AK294095.1 | - | 6 - 7 | 908 | 161 |
AK297195.1 | - | 7 - 8 | 1139 | 161 | |
AK308472.1 | - | 2 - 3 | 646 | 161 | |
AL529385.3 | - | 6 - 7 | 780 | 161 | |
AW954755.1 | - | 2 - 3 | 355 | 161 | |
BC111774.1 | - | 5 - 6 | 1288 | 161 | |
BX357552.2 | - | 3 - 4 | 385 | 161 | |
CX787147.1 | - | 5 - 6 | 639 | 161 | |
U62767.1 | - | 7 - 8 | 868 | 161 |