The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320263.1 | NP_001307192.1 | 7 - 8 | 1296 | 64 |
NM_001320264.1 | NP_001307193.1 | 5 - 6 | 1135 | 64 | |
NM_001320265.1 | NP_001307194.1 | 5 - 6 | 847 | 64 | |
NM_012095.5 | NP_036227.1 | 5 - 6 | 1009 | 64 | |
NM_207012.3 | NP_996895.1 | 6 - 7 | 1135 | 64 | |
NR_135191.1 | 5 - 6 | 1009 | 64 | ||
GenBank mRNA | AF092092.1 | - | 4 - 5 | 675 | 64 |
AK026983.1 | - | 5 - 6 | 744 | 64 | |
AK299352.1 | - | 5 - 6 | 598 | 64 | |
AK315597.1 | - | 5 - 6 | 718 | 64 | |
AL833820.1 | - | 3 - 4 | 314 | 64 | |
BC026232.1 | - | 6 - 7 | 894 | 64 | |
BC067127.1 | - | 5 - 6 | 1105 | 64 |