Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR33
Entrez Gene ID
55339
Gene Name
WD repeat domain 33
Gene Aliases
NET14, WDC146
Chromosome Location
Chr.2: 127704234 - 127811187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018383.4 NP_060853.3 13 - 14 1572 115
XM_005263697.2 XP_005263754.1 13 - 14 1572 115
XM_011511436.1 XP_011509738.1 13 - 14 1601 115
GenBank mRNA AB044749.1 - 13 - 14 1532 115

View Full Product Details