The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018383.4 | NP_060853.3 | 15 - 16 | 1873 | 105 |
XM_005263697.2 | XP_005263754.1 | 15 - 16 | 1873 | 105 | |
XM_011511436.1 | XP_011509738.1 | 15 - 16 | 1902 | 105 | |
GenBank mRNA | AB044749.1 | - | 15 - 16 | 1833 | 105 |