Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3B
Entrez Gene ID
55703
Gene Name
RNA polymerase III subunit B
Gene Aliases
C128, HLD8, INMAP, RPC2
Chromosome Location
Chr.12: 106357658 - 106510198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160708.1 NP_001154180.1 9 - 10 796 110
NM_018082.5 NP_060552.4 9 - 10 949 110
XM_017019621.1 XP_016875110.1 9 - 10 914 110
GenBank mRNA AK122713.1 - 9 - 10 776 110
AK291635.1 - 9 - 10 793 110
AY092084.1 - 9 - 10 780 110
BC046238.1 - 9 - 10 816 110
BX537447.1 - 9 - 10 897 110

View Full Product Details